Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306428.3 | NP_001293357.1 | 1 - 2 | 148 | 100 | |
NM_001383770.1 | 7 - 8 | 1105 | 100 | |||
NM_170806.5 | NP_740789.1 | 6 - 7 | 1182 | 100 | ||
GenBank mRNA | AY008139.1 | - | 6 - 7 | 877 | 100 | |
U56864.1 | - | 6 - 7 | 1094 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |