Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381436.2 | NP_001367838.1 | 11 - 12 | 2215 | 58 | |
NR_131363.1 | 11 - 12 | 2185 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |