Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374886.2 | NP_001361963.1 | 8 - 9 | 2293 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |