Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sem-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170859.6 | NP_740846.1 | 3 - 4 | 308 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |