Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sem-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170859.6 | NP_740846.1 | 6 - 7 | 1093 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |