Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059179.3 | NP_491580.1 | 6 - 7 | 1533 | 80 | |
NM_059180.2 | NP_491581.1 | 6 - 7 | 1412 | 80 | ||
NM_170865.5 | NP_740852.1 | 7 - 8 | 1431 | 80 | ||
NM_182025.7 | NP_871825.1 | 6 - 7 | 1318 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |