Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026412.4 | NP_001021583.1 | 1 - 2 | 100 | 129 | |
GenBank mRNA | AB040992.1 | - | 1 - 2 | 105 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |