Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026412.4 | NP_001021583.1 | 4 - 5 | 679 | 72 | |
GenBank mRNA | AB040992.1 | - | 4 - 5 | 684 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |