Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026412.4 | NP_001021583.1 | 6 - 7 | 1017 | 94 | |
GenBank mRNA | AB040992.1 | - | 6 - 7 | 1022 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |