Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025903.4 | NP_001021074.1 | 1 - 2 | 95 | 81 | |
NM_170901.4 | NP_740891.1 | 1 - 2 | 112 | 81 | ||
GenBank mRNA | AF324832.1 | - | 1 - 2 | 108 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |