Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025903.4 | NP_001021074.1 | 6 - 7 | 706 | 137 | |
NM_170901.4 | NP_740891.1 | 6 - 7 | 723 | 137 | ||
GenBank mRNA | AF324832.1 | - | 6 - 7 | 719 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |