Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026479.4 | NP_001021650.1 | 11 - 12 | 2700 | 95 | |
GenBank mRNA | AJ307058.1 | - | 11 - 12 | 2700 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |