Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026601.6 | NP_001021772.1 | 8 - 9 | 2844 | 131 | |
NM_001383734.1 | 9 - 10 | 2979 | 131 | |||
NM_170927.5 | NP_740922.1 | 8 - 9 | 2844 | 131 | ||
GenBank mRNA | AF332568.1 | - | 8 - 9 | 2838 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |