Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026557.4 | NP_001021728.1 | 5 - 6 | 1114 | 67 | |
NM_001026558.5 | NP_001021729.1 | 4 - 5 | 983 | 67 | ||
GenBank mRNA | AF497832.1 | - | 5 - 6 | 1063 | 67 | |
AF497833.1 | - | 4 - 5 | 1005 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |