Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: catp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129056.5 | NP_001122528.1 | 6 - 7 | 962 | 81 | |
NM_001129057.3 | NP_001122529.1 | 6 - 7 | 961 | 81 | ||
GenBank mRNA | AM419897.1 | - | 6 - 7 | 951 | 81 | |
AM419898.1 | - | 6 - 7 | 951 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |