Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047328.5 | NP_001040793.1 | 2 - 3 | 274 | 62 | |
NM_001047329.4 | NP_001040794.1 | 2 - 3 | 118 | 62 | ||
NM_001381394.1 | NP_001367817.1 | 3 - 4 | 323 | 62 | ||
NM_001381395.2 | NP_001367816.1 | 3 - 4 | 327 | 62 | ||
GenBank mRNA | EU180219.1 | - | 3 - 4 | 307 | 62 |
Gene Symbol: coa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383820.1 | 5 - 6 | 908 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |