Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047328.5 | NP_001040793.1 | 4 - 5 | 876 | 148 | |
NM_001047329.4 | NP_001040794.1 | 4 - 5 | 720 | 148 | ||
NM_001313645.3 | NP_001300574.1 | 1 - 2 | 393 | 148 | ||
NM_001381394.1 | NP_001367817.1 | 5 - 6 | 925 | 148 | ||
NM_001381395.2 | NP_001367816.1 | 5 - 6 | 929 | 148 | ||
NM_170979.6 | NP_740976.1 | 2 - 3 | 831 | 148 | ||
GenBank mRNA | EU180219.1 | - | 5 - 6 | 909 | 148 |
Gene Symbol: coa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383820.1 | 7 - 8 | 1510 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |