Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipla-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047328.5 | NP_001040793.1 | 5 - 6 | 1119 | 126 | |
NM_001047329.4 | NP_001040794.1 | 5 - 6 | 963 | 126 | ||
NM_001313645.3 | NP_001300574.1 | 2 - 3 | 636 | 126 | ||
NM_001381394.1 | NP_001367817.1 | 6 - 7 | 1168 | 126 | ||
NM_001381395.2 | NP_001367816.1 | 6 - 7 | 1172 | 126 | ||
NM_170979.6 | NP_740976.1 | 3 - 4 | 1074 | 126 | ||
GenBank mRNA | EU180219.1 | - | 6 - 7 | 1152 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |