Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170980.4 | NP_740977.1 | 1 - 2 | 189 | 111 | |
NM_171848.6 | NP_740978.1 | 1 - 2 | 180 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |