Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170980.4 | NP_740977.1 | 7 - 8 | 1079 | 90 | |
NM_171848.6 | NP_740978.1 | 7 - 8 | 1070 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |