Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026843.7 | NP_001022014.1 | 11 - 12 | 3210 | 88 | |
NM_001377791.2 | NP_001364725.1 | 5 - 6 | 1944 | 88 | ||
NR_151554.1 | 8 - 9 | 2422 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |