Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026843.7 | NP_001022014.1 | 12 - 13 | 3297 | 105 | |
NM_001377791.2 | NP_001364725.1 | 6 - 7 | 2031 | 105 | ||
NR_151554.1 | 9 - 10 | 2509 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |