Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osta-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171004.7 | NP_741003.1 | 5 - 6 | 784 | 98 | |
NR_136362.1 | 5 - 6 | 784 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |