Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356848.3 | NP_001343765.1 | 2 | 84 | 67 | |
NM_001356849.3 | NP_001343766.1 | 2 | 84 | 67 | ||
NM_001356850.3 | NP_001343767.1 | 1 | 87 | 67 | ||
NM_001356852.3 | NP_001343768.1 | 1 - 2 | 48 | 67 | ||
NM_001383853.1 | 1 - 2 | 44 | 67 | |||
NM_062785.5 | NP_495186.3 | 2 - 3 | 112 | 67 | ||
NM_171007.5 | NP_741006.3 | 2 - 3 | 112 | 67 | ||
NM_171850.7 | NP_741007.2 | 2 - 3 | 131 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |