Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381467.2 | NP_001367422.1 | 8 - 9 | 1653 | 67 | |
NM_062999.9 | NP_495400.2 | 8 - 9 | 1640 | 67 | ||
NM_171014.6 | NP_741015.1 | 8 - 9 | 1637 | 67 | ||
NM_171015.6 | NP_741016.1 | 8 - 9 | 1624 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |