Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351905.4 | NP_001338814.1 | 8 | 1744 | 66 | |
NM_001351913.2 | NP_001338820.1 | 7 - 8 | 1626 | 66 | ||
NM_001381471.2 | NP_001368648.1 | 7 - 8 | 1603 | 66 | ||
NM_001381473.2 | NP_001368649.1 | 6 - 7 | 1538 | 66 | ||
NM_001381475.2 | NP_001368651.1 | 7 - 8 | 1618 | 66 | ||
NM_001381477.2 | NP_001367332.1 | 7 - 8 | 1589 | 66 | ||
NM_001381479.1 | NP_001368654.1 | 6 - 7 | 1530 | 66 | ||
NM_171852.2 | NP_741018.1 | 7 - 8 | 1588 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |