Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171019.3 | NP_741021.1 | 2 - 3 | 218 | 84 | |
NM_171020.5 | NP_741022.1 | 2 - 3 | 208 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |