Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373741.2 | NP_001360471.1 | 19 - 20 | 5539 | 78 | |
NM_001373742.2 | NP_001360472.1 | 19 - 20 | 5536 | 78 | ||
NM_001373743.2 | NP_001360024.1 | 19 - 20 | 5530 | 78 | ||
NM_001373744.2 | NP_001360023.1 | 19 - 20 | 5532 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |