Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171035.6 | NP_741041.1 | 12 - 13 | 1479 | 64 | |
NM_171036.6 | NP_741042.1 | 7 - 8 | 1052 | 64 | ||
GenBank mRNA | LC026472.1 | - | 11 - 12 | 1423 | 64 | |
LC026473.1 | - | 7 - 8 | 937 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |