Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026771.3 | NP_001021942.1 | 26 - 27 | 6188 | 137 | |
NM_001026772.5 | NP_001021943.1 | 14 - 15 | 4141 | 137 | ||
NM_001026773.5 | NP_001021944.1 | 8 - 9 | 3299 | 137 | ||
NM_001267341.4 | NP_001254270.1 | 23 - 24 | 5643 | 137 | ||
NM_001276741.3 | NP_001263670.1 | 27 - 28 | 6260 | 137 | ||
NM_001276742.3 | NP_001263671.1 | 27 - 28 | 6392 | 137 | ||
NM_001276743.3 | NP_001263672.1 | 26 - 27 | 6320 | 137 | ||
NM_001276744.4 | NP_001263673.1 | 8 - 9 | 3947 | 137 | ||
NM_001276745.3 | NP_001263674.1 | 6 - 7 | 2897 | 137 | ||
NM_001377858.3 | NP_001364824.1 | 27 - 28 | 5930 | 137 | ||
NM_001377859.2 | NP_001364823.1 | 26 - 27 | 5858 | 137 | ||
NM_001377860.2 | NP_001364658.1 | 24 - 25 | 5675 | 137 | ||
NM_001377861.2 | NP_001364506.1 | 24 - 25 | 5383 | 137 | ||
NM_001377862.2 | NP_001364507.1 | 23 - 24 | 5273 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |