Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026998.5 | NP_001022169.1 | 13 - 14 | 1861 | 78 | |
NM_001026999.8 | NP_001022170.1 | 13 - 14 | 1861 | 78 | ||
NM_001027000.6 | NP_001022171.1 | 10 - 11 | 1656 | 78 | ||
NM_001027001.5 | NP_001022172.1 | 5 - 6 | 601 | 78 | ||
NM_001267342.3 | NP_001254271.1 | 13 - 14 | 1861 | 78 | ||
NM_001267343.3 | NP_001254272.1 | 13 - 14 | 1861 | 78 | ||
NM_001267344.3 | NP_001254273.1 | 6 - 7 | 937 | 78 | ||
NM_171039.3 | NP_741045.1 | 13 - 14 | 1902 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |