Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171096.6 | NP_741109.1 | 1 - 2 | 123 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |