Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171109.4 | NP_741122.2 | 2 - 3 | 346 | 87 | |
NM_171863.4 | NP_741123.2 | 2 - 3 | 340 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |