Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027596.4 | NP_001022767.1 | 3 - 4 | 776 | 120 | |
NM_001379744.1 | NP_001367437.1 | 3 - 4 | 781 | 120 | ||
GenBank mRNA | U32275.1 | - | 3 - 4 | 784 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |