Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027422.4 | NP_001022593.1 | 1 - 2 | 198 | 78 | |
NM_001027423.6 | NP_001022594.1 | 1 - 2 | 210 | 78 | ||
NM_001027425.5 | NP_001022596.1 | 1 - 2 | 155 | 78 | ||
NM_001027426.4 | NP_001022597.1 | 1 - 2 | 212 | 78 | ||
NM_001379757.2 | NP_001368076.1 | 3 - 4 | 334 | 78 | ||
GenBank mRNA | AF272738.1 | - | 1 - 2 | 155 | 78 | |
AY053364.1 | - | 1 - 2 | 155 | 78 | ||
AY053365.1 | - | 1 - 2 | 155 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |