Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379763.1 | NP_001367440.1 | 5 - 6 | 2478 | 74 | |
NM_171169.6 | NP_741197.1 | 4 - 5 | 1357 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |