Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379776.1 | NP_001366805.1 | 6 - 7 | 564 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |