Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flap-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171178.7 | NP_741207.1 | 5 - 6 | 561 | 76 | |
NM_171879.6 | NP_741208.1 | 6 - 7 | 611 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |