Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171180.5 | NP_741210.3 | 4 - 5 | 884 | 68 | |
NM_171880.6 | NP_741211.3 | 4 - 5 | 886 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |