Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025993.4 | NP_001021164.1 | 2 - 3 | 360 | 75 | |
GenBank mRNA | L19248.1 | - | 1 - 2 | 405 | 75 | |
L19639.1 | - | 2 - 3 | 343 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |