Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066372.7 | NP_498773.3 | 14 - 15 | 4224 | 77 | |
NM_171207.8 | NP_741245.3 | 13 - 14 | 3975 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |