Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 14 - 15 | 2550 | 66 | |
NM_001129275.4 | NP_001122747.1 | 4 - 5 | 653 | 66 | ||
NM_001129276.3 | NP_001122748.1 | 15 - 16 | 2634 | 66 | ||
NM_001268079.4 | NP_001255008.1 | 15 - 16 | 2649 | 66 | ||
NM_001268080.3 | NP_001255009.1 | 10 - 11 | 1739 | 66 | ||
NM_001306679.3 | NP_001293608.1 | 8 - 9 | 1535 | 66 | ||
NM_001306687.3 | NP_001293616.1 | 9 - 10 | 1553 | 66 | ||
NM_001306688.3 | NP_001293617.1 | 5 - 6 | 737 | 66 | ||
NM_001382965.1 | 17 - 18 | 2955 | 66 | |||
NM_171221.7 | NP_741263.3 | 16 - 17 | 2641 | 66 | ||
GenBank mRNA | AF424978.1 | - | 14 - 15 | 2486 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |