Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027709.3 | NP_001022880.2 | 16 - 17 | 3170 | 63 | |
GenBank mRNA | M93256.1 | - | 16 - 17 | 3185 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |