Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elo-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268181.3 | NP_001255110.1 | 2 - 3 | 182 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |