Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027443.5 | NP_001022614.1 | 2 - 3 | 225 | 109 | |
NM_001379980.1 | NP_001366988.1 | 2 - 3 | 227 | 109 | ||
GenBank mRNA | AB008842.1 | - | 2 - 3 | 235 | 109 | |
AB008843.1 | - | 2 - 3 | 235 | 109 | ||
AB008844.1 | - | 2 - 3 | 235 | 109 | ||
AF047885.1 | - | 2 - 3 | 237 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |