Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171270.2 | NP_741320.1 | 2 - 3 | 677 | 75 | |
NM_171271.6 | NP_741321.1 | 3 - 4 | 1956 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |