Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380065.1 | NP_001367952.1 | 5 - 6 | 1051 | 64 | |
NM_001380066.2 | NP_001367951.1 | 5 - 6 | 1070 | 64 | ||
GenBank mRNA | AF192746.1 | - | 3 - 4 | 835 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |