Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028392.7 | NP_001023563.1 | 5 - 6 | 459 | 74 | |
NM_001307684.3 | NP_001294613.1 | 3 - 4 | 201 | 74 | ||
NM_001383055.1 | 3 - 4 | 1269 | 74 | |||
GenBank mRNA | AF239999.2 | - | 4 - 5 | 409 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |