Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380163.1 | NP_001367796.1 | 4 - 5 | 601 | 103 | |
NM_001380164.2 | NP_001367795.1 | 4 - 5 | 615 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |