Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028204.4 | NP_001023375.1 | 5 - 6 | 1851 | 103 | |
NM_171344.5 | NP_741403.2 | 6 - 7 | 2026 | 103 | ||
GenBank mRNA | AF166274.1 | - | 5 - 6 | 1858 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |